Learn More
World Precision Instrument Reusable Biopsy Punch, Various Sizes-504533
SDP
Catalog No. NC1060510
Encompass
Our reusable biopsy punches are multi-purpose sampling tools designed with a retractable cutting cannula. They are easy to operate, efficient, safe, fast and precise. Our punches consists of a razor sharp stainless steel cutting tip. It is designed to cut, retrieve and store the cored samples from a source materials. The source could be skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The biopsy punches are recommended as a DNA sampling tool and are also ideal for forensic use and laboratory sampling applications. The punches come sterile and are individually wrapped. They were sterilized using ethylene oxide.
Catalog No. NC1060510 Supplier World Precision Instrument Supplier No. 504533
Our reusable biopsy punches are multi-purpose sampling tools designed with a retractable cutting cannula. They are easy to operate, efficient, safe, fast and precise. Our punches consists of a razor sharp stainless steel cutting tip. It is designed to cut, retrieve and store the cored samples from a source materials. The source could be skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The biopsy punches are recommended as a DNA sampling tool and are also ideal for forensic use and laboratory sampling applications. The punches come sterile and are individually wrapped. They were sterilized using ethylene oxide.