Learn More
Med Vet International Miltex Sterile Disposable Biopsy Punch, 3.5mm, 50/Box
Catalog No. 502829564
Encompass_Preferred
The Miltex 33-33 3.5mm sterile disposable biopsy dermal punch has a blade made of OR grade German stainless steel. Features: Seamless, stainless steel, razor-sharp cutting edges eliminate torn or pulled biopsies Ribbed handle provides comfort and control Convenience of disposables, single-use Sizes clearly marked on each punch Individual color-coded blister packs Sterile, 5-year shelf life Each box contains 50 punches.
Catalog No. 50-282-9564 Supplier Med Vet International Supplier No. MIL3333
The Miltex 33-33 3.5mm sterile disposable biopsy dermal punch has a blade made of OR grade German stainless steel. Features: Seamless, stainless steel, razor-sharp cutting edges eliminate torn or pulled biopsies Ribbed handle provides comfort and control Convenience of disposables, single-use Sizes clearly marked on each punch Individual color-coded blister packs Sterile, 5-year shelf life Each box contains 50 punches.