Learn More
Med Vet International Dynarex Biopsy Punches, Mix Box: 2, 3, 4, 5 And 6 mm, 5 Each Size, 25/Box
Catalog No. 502829181
Encompass_Preferred
Dynarex Biopsy Punches are premium-quality medical instruments designed to accurately and effectively cut and retrieve tissue samples for laboratory testing. Each sterile Biopsy Punch features a durable, ribbed handle for improved control and a seamless, sharp stainless-steel blade for improved efficiency. Dynarex Biopsy Punches are available in a variety of color-coded sizes suitable for a range of procedures. Dynarex Biopsy Punches feature a seamless stainless-steel blade with a durable, ribbed handle, designed to provide greater comfort and control when obtaining tissue samples.
Catalog No. 50-282-9181 Supplier Med Vet International Supplier No. 4097
Dynarex Biopsy Punches are premium-quality medical instruments designed to accurately and effectively cut and retrieve tissue samples for laboratory testing. Each sterile Biopsy Punch features a durable, ribbed handle for improved control and a seamless, sharp stainless-steel blade for improved efficiency. Dynarex Biopsy Punches are available in a variety of color-coded sizes suitable for a range of procedures. Dynarex Biopsy Punches feature a seamless stainless-steel blade with a durable, ribbed handle, designed to provide greater comfort and control when obtaining tissue samples.