Learn More
Ihc World LLC BIOPSY PUNCH NEEDLES W PLUGR4
SDP
Catalog No. NC0726265
Encompass_Preferred
Biopsy Punch Needles with Plunger 3mm, 4 pcs/pack. The Biopsy Punch Needles with Plunger are used for building tissue microarray blocks. Features: Effortless ejection of specimens. Seamless, stainless steel, razor-sharp cutting edges. Ribbed handle provides comfort and control. Disposable and reusable. Sizes clearly marked on each punch. Diameter: 3mm
Catalog No. NC0726265 Supplier Ihc World LLC Supplier No. IW1253
Biopsy Punch Needles with Plunger 3mm, 4 pcs/pack. The Biopsy Punch Needles with Plunger are used for building tissue microarray blocks. Features: Effortless ejection of specimens. Seamless, stainless steel, razor-sharp cutting edges. Ribbed handle provides comfort and control. Disposable and reusable. Sizes clearly marked on each punch. Diameter: 3mm