Learn More
Ihc World LLC BIOPSY PUNCH NEEDLES W/PLUNGER
SDP
Catalog No. NC9875630
Encompass_Preferred
The Biopsy Punch Needles with Plunger are used for building tissue microarray blocks. Features:
  • Effortless ejection of specimens.
  • Seamless, stainless steel, razor-sharp cutting edges.
  • Ribbed handle provides comfort and
Catalog No. NC9875630 Supplier Ihc World LLC Supplier No. IW1254
The Biopsy Punch Needles with Plunger are used for building tissue microarray blocks. Features:
  • Effortless ejection of specimens.
  • Seamless, stainless steel, razor-sharp cutting edges.
  • Ribbed handle provides comfort and