Learn More
Alkali Scientific ASI™ Biopsy Sponge Pads for Cassettes, Foam, Blue, 30.2mm x 25.4mm x 2mm, 1000/Pack, 10,000/Case
SDP
Catalog No. 503026541
Encompass_Preferred
These blue biopsy pads are expertly crafted from uniform, polyester reticulated open-cell foam, providing an ideal solution for secure and efficient biopsy processing. The open-cell structure allows fluids to flow freely and thoroughly around the tissue, ensuring safe and complete specimen preservation while minimizing handling risks. The thin 2mm design is thoughtfully engineered to reduce pressure on unfixed specimens, effectively lowering the likelihood of tissue sticking to the pads during embedding. This gentle yet secure approach helps maintain specimen integrity, making these pads a trusted choice for precise histological work. Proudly made in the USA, these biopsy pads ensure the proper orientation of small biopsy tissues, aiding in accurate diagnostic results. Whether for high-throughput labs or small-scale operations, these pads deliver reliable performance and enhanced efficiency.
Catalog No. 50-302-6541 Supplier Alkali Scientific Supplier No. BP01CS
These blue biopsy pads are expertly crafted from uniform, polyester reticulated open-cell foam, providing an ideal solution for secure and efficient biopsy processing. The open-cell structure allows fluids to flow freely and thoroughly around the tissue, ensuring safe and complete specimen preservation while minimizing handling risks. The thin 2mm design is thoughtfully engineered to reduce pressure on unfixed specimens, effectively lowering the likelihood of tissue sticking to the pads during embedding. This gentle yet secure approach helps maintain specimen integrity, making these pads a trusted choice for precise histological work. Proudly made in the USA, these biopsy pads ensure the proper orientation of small biopsy tissues, aiding in accurate diagnostic results. Whether for high-throughput labs or small-scale operations, these pads deliver reliable performance and enhanced efficiency.