Learn More
Newcomer Supply Biopsy Sponge Pads for Cassettes - Blue, 10,000/case
SDP
Catalog No. NC0259333
Encompass
These pads are used to hold biopsies in place and prevent them from being lost during processing. They are made of a polyester urethane foam which is always verified for consistency throughout to achieve optimum solvent flow. This pad will fit the majority of tissue cassettes for this purpose.  Will resist temps. from -40°C to 121°C. Dimensions: 30.2 x 25.4 x 2mm
Catalog No. NC0259333 Supplier Newcomer Supply Supplier No. 5106A
These pads are used to hold biopsies in place and prevent them from being lost during processing. They are made of a polyester urethane foam which is always verified for consistency throughout to achieve optimum solvent flow. This pad will fit the majority of tissue cassettes for this purpose.  Will resist temps. from -40°C to 121°C. Dimensions: 30.2 x 25.4 x 2mm