Learn More
LabScientific Uniform polyester reticulated open cell foam biopsy sponges for biopsy tissue. Smooth flow, keeps specimen secure. Assures orientation of specimen. 1000/bag.
Catalog No. NC0631506
Encompass
Uniform polyester reticulated open cell foam Biopsy Pads allow fluids to transfer safely and thoroughly around the tissue while holding the specimen secure. The thin 2mm design helps eliminate pressure from being applied to the unfixed specimen, reducing the risk of tissue sticking to the pads during embedding. Made In the USA. Assures orientation of small biopsy tissue. Uniform open cell polyester foam. Dimensions: 30.2mm x 25mm x 2mm. 1000/bag
Catalog No. NC0631506 Supplier LabScientific Supplier No. HBS
Uniform polyester reticulated open cell foam Biopsy Pads allow fluids to transfer safely and thoroughly around the tissue while holding the specimen secure. The thin 2mm design helps eliminate pressure from being applied to the unfixed specimen, reducing the risk of tissue sticking to the pads during embedding. Made In the USA. Assures orientation of small biopsy tissue. Uniform open cell polyester foam. Dimensions: 30.2mm x 25mm x 2mm. 1000/bag