Learn More
Bioptechs™ Delta T Culture Dishes
Catalog No. p-4703886
Encompass
Change view
Click to view available options
Color:
Black
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12-071-210 Clear
12-071-211 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-071-210 Supplier Bioptechs™ Supplier No. 04200417C50
Only null left
Add to Cart
Add to Cart
For used in fluorescence, DIC, and confocal modes of microscopy, or anywhere that highest image quality is desirable
  • Disposable plastic culture dish with the high optical compatibility of glass
  • Contain self heating ITO coating, which provides first surface heat transfer directly to cells when using Delta T System
  • Injection molded polystyrene sidewalls and lid, coverglass bottom
  • Highest quality German Desag 263, low alkaline coverglass for optimum resolution
  • 35mm O.D. and 23mm central aperture with tapered sidewalls and low profile (6mm in height) to reduce dead-volume and allow better access for microinjection and micromanipulation
  • Glass thicknesses of 0.17mm
  • Available in clear and black (for fluorescence applications)
  • Numerous accessories specific to these dishes allow for specialized applications including - media perfusion, gas perfusion, sample positioning, heating and cooling
  • Optional stage adapters available to fit all major brands of microscopes, which custom stage adapters available
  • Optional culturing cylinders may be used to barricade cells or suspended specimens in a dish; or to restrict and concentrate growth and location of cells plated on coverslip without use of grease
Specifications
Color Clear