Learn More
BioSpec Products BIOPULVERIZER, CAPACITY 0.1-1G
SDP
Catalog No. NC0400959
Encompass_Preferred
BioPulverizers quickly fragment hard frozen tissue samples into pieces the size of grains of sand or smaller. capacity 0.1-1 g
Catalog No. NC0400959 Supplier BioSpec Products Supplier No. 59013N
BioPulverizers quickly fragment hard frozen tissue samples into pieces the size of grains of sand or smaller. capacity 0.1-1 g