Learn More
BioSpec Products BIOPULVERIZER, CAPACITY 1-10G
SDP
Catalog No. NC0283690
Encompass_Preferred
BioPulverizers quickly fragment hard frozen tissue samples into pieces the size of grains of sand or smaller. capacity 1-10 g
Catalog No. NC0283690 Supplier BioSpec Products Supplier No. 59014N
BioPulverizers quickly fragment hard frozen tissue samples into pieces the size of grains of sand or smaller. capacity 1-10 g