Learn More
BioSpec Products BIOPULVERIZER,MULTI 12WELL
SDP
Catalog No. NC1340586
Encompass_Preferred
Multisample Biopulverizer, 12 wells with 10-100mg per well. Model 59012MS
Catalog No. NC1340586 Supplier BioSpec Products Supplier No. 59012MS
Multisample Biopulverizer, 12 wells with 10-100mg per well. Model 59012MS