Learn More
Ted Pella Inc Biopunch, ID 6.0mm, OD 6.47mm, 5/pk
SDP
Catalog No. NC0669799
Encompass
The Biopunch is a coring tool designed to core, punch and retrieve samples from thin or soft substrate source materials such as tissue, gels, paper, cloth, leaves, paint chips and films.
Catalog No. NC0669799 Supplier Ted Pella Inc Supplier No. 1511160
The Biopunch is a coring tool designed to core, punch and retrieve samples from thin or soft substrate source materials such as tissue, gels, paper, cloth, leaves, paint chips and films.