Learn More
Diversified Biotech Inc Bio-Pure Alcohol Wipes, 480/case
Catalog No. NC1642041
Encompass
Proprietary fabric is formulated to easily break down over time. Re-sealable and flexible packaging has a low carbon footprint. 70% IPA/30% H2O mixture evaporates completely. Wipes won't scratch surfaces of optics. Strong fabric won't fall apart. Idea use: Clean lab surfaces, equipment, optical parts and cuvettes.
This item is not returnable. View return policy
Catalog No. NC1642041 Supplier Diversified Biotech Inc Supplier No. BPWP1000CS
Proprietary fabric is formulated to easily break down over time. Re-sealable and flexible packaging has a low carbon footprint. 70% IPA/30% H2O mixture evaporates completely. Wipes won't scratch surfaces of optics. Strong fabric won't fall apart. Idea use: Clean lab surfaces, equipment, optical parts and cuvettes.