Learn More
Higgins Analytical Inc BIOPURESPE MACRO 96-WELL,100MG
Catalog No. NC1962840
Encompass
BioPureSPN Macro-96-Well, PROTO 300 C18, 35-350µg max capacity, 0.2ml, 100mg, 300Å
Catalog No. NC1962840 Supplier Higgins Analytical Inc Supplier No. HNS S18VL
BioPureSPN Macro-96-Well, PROTO 300 C18, 35-350µg max capacity, 0.2ml, 100mg, 300Å