Learn More
Higgins Analytical Inc BioPureSPN Mini, PROTO 300 C18, 90bx
Catalog No. NC1678001
Encompass
BioPureSPN Mini, PROTO 300 C18 (Green) 7-70µg capacity, 0.2ml, 20mg, 300Å, 90/bx
Catalog No. NC1678001 Supplier Higgins Analytical Inc Supplier No. HUM S18V
BioPureSPN Mini, PROTO 300 C18 (Green) 7-70µg capacity, 0.2ml, 20mg, 300Å, 90/bx