Learn More
QA Bio Inc BioQuant Quantitative Man-8 Standard
Catalog No. 502236502
Encompass
The BioQuant quantitative Man8 glycan is a purified and quantified glycan standard which can be used as an internal standard and positive control for quantitative analysis (e.g. monosaccharide analysis). • 10 µg • The amount of BioQuant quantitative Man8 glycan to be dispensed per vial is determined by quantitative Nuclear Magnetic Resonance (qNMR) of the bulk glycan stock. Once dispensed the amount of glycan per vial is determined by monosaccharide analysis.• Glycan Purity determined as > 90% by UHPLC • Monoisotopic mass: 1721.6 [M+H]+
This item is not returnable. View return policy
Catalog No. 50-223-6502 Supplier QA Bio Inc Supplier No. BQCNMAN810U
The BioQuant quantitative Man8 glycan is a purified and quantified glycan standard which can be used as an internal standard and positive control for quantitative analysis (e.g. monosaccharide analysis). • 10 µg • The amount of BioQuant quantitative Man8 glycan to be dispensed per vial is determined by quantitative Nuclear Magnetic Resonance (qNMR) of the bulk glycan stock. Once dispensed the amount of glycan per vial is determined by monosaccharide analysis.• Glycan Purity determined as > 90% by UHPLC • Monoisotopic mass: 1721.6 [M+H]+