Learn More
BioQuest™ Inflatable Lungs Kit
SCI_ED
Catalog No. s801131
Encompass
Change view
Click to view available options
:
Inflatable Lungs Kit
Catalog No.
S801131 Inflatable Lungs Kit
1 options
Choking Hazard: Small Parts. Not for children under 3 years.
Catalog No. S801131 Supplier Nasco Supplier No. LS03765DI
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Choking Hazard: Small Parts. Not for children under 3 years.
Includes
Inflation rack and tray, air pump, one pair of reusable and inflatable swine lungs, one section of dried swine lung, a teacher's guide, and a 2 qt. container for storing the lungs
Dramatic demonstration kit provides an impressive, unforgettable, hands-on demonstration of the texture and fragility of the mammalian lung.
  • Demonstrates that real lungs are not like rubber balloons in a bell jar
  • Demonstrates the touch and feel of a lung that is very similar to a human lung
  • Suitable for elementary, secondary and college classes in health, biology, anatomy and physical education
  • Includes supplemental information about lung cancer, other smoking-induced diseases and air pollutants
  • Teacher's guide emphasizes the handling, use, and storage of the reusable swine lungs and includes a detailed illustration of internal anatomy
  • Kit includes swine lungs that have been specially preserved by the BioFlex process
  • Students can see and learn how lungs work by inflating and deflating them with a pump
  • Also includes a section of dried lung to show internal anatomy structures including arteries, veins, bronchioles, alveoli and bronchi
Specifications
For Use With (Application) Student can see and learn how lungs, that are very comparable with their own, work by inflating and deflating them with a pump
Includes Inflation rack and tray, air pump, one pair of reusable and inflatable swine lungs, one section of dried swine lung, teacher's guide and 2 qt. container for storing the lungs, detailed illustration of internal anatomy, supplemental information about lung
Product Type Inflatable Lungs Kit