Learn More
Bio Rad Laboratories BIORAD DC PROTEIN ASSAY KIT II
Catalog No. NC9332008
Encompass
Detergent-compatible colorimetric assay kit, includes 250 ml alkaline copper tartrate, 2 L dilute Folin reagent, 5 ml surfactant solution, BSA standard; 450 assays
Catalog No. NC9332008 Supplier Bio Rad Laboratories Supplier No. 5000112
Detergent-compatible colorimetric assay kit, includes 250 ml alkaline copper tartrate, 2 L dilute Folin reagent, 5 ml surfactant solution, BSA standard; 450 assays