Learn More
Kuhner Shaker Inc BIOREACTOR
Catalog No. 502569957
Encompass
--PLEASE CONTACT KUHNER SHAKER (usoffice@kuhner.com) FOR ADDITIONAL DETAILS AND TO SET UP AN ONSITE DEMO-- 10L Orbitally Shaken Bioreactor SB10-X. The SB10-X is a shaken benchtop bioreactor system for the cultivation of mammalian and plant cells in a single-use 3D bag (working volume from 3 to 12L). No sparging or impeller needed, allowing for seamless scale-up of shake flask processes and elimination of elaborate cleaning and sterilization protocols. Fully functional bioreactor with gas overlay, live pH/DO monitoring and control and multiple ports for all liquid feeding/harvesting. Reactor Dimensions incl. shaker: WxDxH: 613 x 719 x 775 mm. Control Unit Dimensions: WxDxH: 303 x 673 x 582 mm. Reactor weight incl. shaker:75 kg; Contol unit Weight: 30 kg; Shaking diameter: 50mm (Orbital); Speed: max. 140 rpm +/- 0.1 rpm; Temperature range: Ambient +5 - 45 °C (No Cooling)
Catalog No. 50-256-9957 Supplier Kuhner Shaker Inc Supplier No. SMX7600
Add to Cart
Add to Cart
Request a Quote
--PLEASE CONTACT KUHNER SHAKER (usoffice@kuhner.com) FOR ADDITIONAL DETAILS AND TO SET UP AN ONSITE DEMO-- 10L Orbitally Shaken Bioreactor SB10-X. The SB10-X is a shaken benchtop bioreactor system for the cultivation of mammalian and plant cells in a single-use 3D bag (working volume from 3 to 12L). No sparging or impeller needed, allowing for seamless scale-up of shake flask processes and elimination of elaborate cleaning and sterilization protocols. Fully functional bioreactor with gas overlay, live pH/DO monitoring and control and multiple ports for all liquid feeding/harvesting. Reactor Dimensions incl. shaker: WxDxH: 613 x 719 x 775 mm. Control Unit Dimensions: WxDxH: 303 x 673 x 582 mm. Reactor weight incl. shaker:75 kg; Contol unit Weight: 30 kg; Shaking diameter: 50mm (Orbital); Speed: max. 140 rpm +/- 0.1 rpm; Temperature range: Ambient +5 - 45 °C (No Cooling)