Learn More
Reprocell BIOREACTOR CONTROLLER & MOTOR
Catalog No. NC1813142
Encompass
This controller works with the Able Bioreactor Magnetic Stir System Base (5 mL; Cat. No. ABBWBP05N0S-6) or the Able Bioreactor Stir System Base (30 mL, 100 mL: Cat. No. ABBWBP03N0S-6) to provide consistent, heat-free stirring of disposable bioreactors. REPROCELL mfg# ABBWDW-1013
Catalog No. NC1813142 Supplier Reprocell Supplier No. ABBWDW1013
This controller works with the Able Bioreactor Magnetic Stir System Base (5 mL; Cat. No. ABBWBP05N0S-6) or the Able Bioreactor Stir System Base (30 mL, 100 mL: Cat. No. ABBWBP03N0S-6) to provide consistent, heat-free stirring of disposable bioreactors. REPROCELL mfg# ABBWDW-1013