Learn More
Thermo Scientific™ Bioreactor Probe Assembly for use with Single-Use Bioreactors
Shop All Thermo Scientific Products

Description
The Thermo Scientific™ Bioreactor Probe Assembly (non-sterile) is used to package the pH and DO probes for sterilization and to aseptically connect them to the BPCs for Thermo Scientific™ HyPerforma™ Single-Use Bioreactors.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Silicone |
| For Use With | DynaDrive™ Single-use Bioreactors, HyPerforma™ Single-use Bioreactors |
| Sterility | Non-sterile |
| Includes | Kleenpak Connector |
| Color | Clear |
| Product Type | Probe Assembly |
| Quantity | 10 Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.