Learn More
Bellco Glass, Inc. Bioreactor Vessel, 3L
SDP
Catalog No. 501946466
Encompass_Preferred
Round bottom bioreactor vessel is manufactured of type 1 class a Borosilicate glass. Graduations are 500ml to 2500ml every 500ml. Vessel dimensions are 145mm od x 265mm oah. A full range of accessories are available for this vessel. 3L.. MADE IN THE USA
Catalog No. 50-194-6466 Supplier Bellco Glass, Inc. Supplier No. 790910003
Round bottom bioreactor vessel is manufactured of type 1 class a Borosilicate glass. Graduations are 500ml to 2500ml every 500ml. Vessel dimensions are 145mm od x 265mm oah. A full range of accessories are available for this vessel. 3L.. MADE IN THE USA