Learn More
Nanocomposix BioReady Gold Nanoshells – Carboxyl - 150 nm, 20 OD in aqueous 0.02 mM potassium carbonate, 30 mL
SDP
Catalog No. NC1579992
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.150 nm gold nanoshells for covalent conjugation combine strong plasmonic optical properties with readily biofunctionalized surface chemistry. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.BioReady Gold Nanoshells enable enhanced sensitivity in lateral flow assays (LFA) due to their intense optical signal. Integration of these nanoparticles into LFAs can provide increased signal-to-noise ratios and lowered limits of detection for your target of interest.BioReady Gold Nanoshells have carboxyl (lipoic acid) covalently bound to the metal surface. Antibodies and other amine-containing biomolecules efficiently bind to carboxyl gold nanoshells using EDC-NHS chemistry to make highly stable conjugates.
Catalog No. NC1579992 Supplier Nanocomposix Supplier No. GSXR15030M
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.150 nm gold nanoshells for covalent conjugation combine strong plasmonic optical properties with readily biofunctionalized surface chemistry. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.BioReady Gold Nanoshells enable enhanced sensitivity in lateral flow assays (LFA) due to their intense optical signal. Integration of these nanoparticles into LFAs can provide increased signal-to-noise ratios and lowered limits of detection for your target of interest.BioReady Gold Nanoshells have carboxyl (lipoic acid) covalently bound to the metal surface. Antibodies and other amine-containing biomolecules efficiently bind to carboxyl gold nanoshells using EDC-NHS chemistry to make highly stable conjugates.