Learn More
Nanocomposix BioReady Gold Nanoshells – Citrate - 150 nm, 20 OD in aqueous 0.2 mM sodium citrate, 5 mL
SDP
Catalog No. NC2171822
Encompass_Preferred
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.150 nm gold nanoshells for passive adsorption combine strong plasmonic optical properties with surface chemistry designed for passive conjugation. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.BioReady Gold Nanoshells enable enhanced sensitivity in lateral flow assays (LFA) due to their intense optical signal. Integration of these nanoparticles into LFAs can provide increased signal-to-noise ratios and lowered limits of detection for your target of interest.
This item is not returnable. View return policy
Catalog No. NC2171822 Supplier Nanocomposix Supplier No. GSCR1505M
As a global leader in gold nanoparticle production, nanoComposix specializes in providing high-purity and monodisperse nanomaterials -- delivering the quality you need for consistent results.150 nm gold nanoshells for passive adsorption combine strong plasmonic optical properties with surface chemistry designed for passive conjugation. These nanoparticles are used in applications such as immunodiagnostics, lateral flow assays, nanomedicine, drug delivery, biosensors, and more.BioReady Gold Nanoshells enable enhanced sensitivity in lateral flow assays (LFA) due to their intense optical signal. Integration of these nanoparticles into LFAs can provide increased signal-to-noise ratios and lowered limits of detection for your target of interest.