Learn More
MSC Clift Industries Surface Cleaner, 275 Gallon Tote, Oil & Grease Remediation
Catalog No. 502837619
Encompass
BioRem-2000 Surface Cleaner is an amber liquid designed to remediate oil, grease, and other hydrocarbons from hard surfaces. This product utilizes microbial technology to effectively eliminate contaminants, restoring surfaces to a clean, safe, and dry state. It is non-hazardous and has a neutral pH, making it suitable for various cleanup and bioremediation processes.
  • Specializes in eradicating hydrocarbons from hard surfaces, water, and soil
  • Can be easily used with mops, commercial floor scrubbers, pressure washers, and other equipment
  • Non-hazardous, has a neutral pH, and exhibits low toxicity to plant, animal, and marine life
  • Provides cost-effective cleanup for hard surfaces and contaminated sites
  • Allows for significant reduction and eradication of petroleum-based contaminants at greater speed and lower risk
Catalog No. 50-283-7619 Supplier MSC Supplier No. 48380109
BioRem-2000 Surface Cleaner is an amber liquid designed to remediate oil, grease, and other hydrocarbons from hard surfaces. This product utilizes microbial technology to effectively eliminate contaminants, restoring surfaces to a clean, safe, and dry state. It is non-hazardous and has a neutral pH, making it suitable for various cleanup and bioremediation processes.
  • Specializes in eradicating hydrocarbons from hard surfaces, water, and soil
  • Can be easily used with mops, commercial floor scrubbers, pressure washers, and other equipment
  • Non-hazardous, has a neutral pH, and exhibits low toxicity to plant, animal, and marine life
  • Provides cost-effective cleanup for hard surfaces and contaminated sites
  • Allows for significant reduction and eradication of petroleum-based contaminants at greater speed and lower risk