Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Bioremediation by Oil-eating Bacteria
SDPSCI_ED

Catalog No. s68637 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S68637 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68637 Supplier Edvotek™ Supplier No. 956
Only null left
Add to Cart
Add to Cart

Includes

instructions, oil-eating microbes (powder), growth media, petri plates, 1N HCl, 1M NaOH, tetrazolium indicator powder, pH paper, and pipets

Requires

  • Shaking incubation oven or stirrer and stir bars
  • Autoclave
  • Vegetable oil and other food oils

Examine the process by which compounds are broken down by microbes.

Bioremediation enhances this natural process to mitigate the effects of oil spills or other pollution events.

  • Grow a mixture of oil-eating bacteria
  • Demonstrate how their enzymes digest hydrocarbon molecules, promoting degradation

Specifications

Grade 9 to Undergraduate
For Use With (Application) To demonstrate bioremediation by oil eating bacteria
Includes Instructions, oil eating bacteria, growth medium, pipets
Time Range 50 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Simulation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.