Learn More
Waters Corp Waters BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 1 mm × 150 mm
Catalog No. NC1732449 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Column is designed for high-performance reverse phase protein separation and analysis. It utilizes 2.7 μm superficially-porous-silica particles with a 450Å pore diameter and a proprietary polyphenyl bonding process for LC or LC-MS of intact IgG monoclonal antibodies and their fragments.
  • Qualified using a gradient separation of the mAb Subunit Standard for reproducible performance.
  • Ideal for intact protein and mAb subunit analysis, protein analysis, ADC characterization, AAV, and antibody conjugates.
  • Improves component resolution, peak capacity, and minimizes injection-to-injection carryover.
  • Compatible with UPLC, UHPLC, and HPLC platforms.
  • Allows lower temperature operation to minimize on-column sample degradation.
  • Manufactured in a cGMP, ISO 9001-certified facility using pure reagents.
Catalog No. NC1732449 Supplier Waters Corp Supplier No. 186009017
The BioResolve RP mAb Polyphenyl Column is designed for high-performance reverse phase protein separation and analysis. It utilizes 2.7 μm superficially-porous-silica particles with a 450Å pore diameter and a proprietary polyphenyl bonding process for LC or LC-MS of intact IgG monoclonal antibodies and their fragments.
  • Qualified using a gradient separation of the mAb Subunit Standard for reproducible performance.
  • Ideal for intact protein and mAb subunit analysis, protein analysis, ADC characterization, AAV, and antibody conjugates.
  • Improves component resolution, peak capacity, and minimizes injection-to-injection carryover.
  • Compatible with UPLC, UHPLC, and HPLC platforms.
  • Allows lower temperature operation to minimize on-column sample degradation.
  • Manufactured in a cGMP, ISO 9001-certified facility using pure reagents.