Learn More
Waters Corp Waters BioResolve CX pH Concentrate, 10X Buffer, pH 5.0 to 10.2 Gradient
Catalog No. 502038895 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 10X buffer concentrate is optimized for creating a pH gradient for monoclonal antibody (mAb) charge variant separations. It is designed for use with BioResolve SCX columns.
  • 10X buffer concentrate
  • Optimized for creating a pH 5.0 to pH 10.2 gradient
  • Designed for use in monoclonal antibody (mAb) charge variant separations
  • Developed alongside BioResolve SCX columns
Catalog No. 50-203-8895 Supplier Waters Corp Supplier No. 186009063
This 10X buffer concentrate is optimized for creating a pH gradient for monoclonal antibody (mAb) charge variant separations. It is designed for use with BioResolve SCX columns.
  • 10X buffer concentrate
  • Optimized for creating a pH 5.0 to pH 10.2 gradient
  • Designed for use in monoclonal antibody (mAb) charge variant separations
  • Developed alongside BioResolve SCX columns