Learn More
Waters Corp Waters Corporation pH Concentrate, 10X, pH 10.2
Catalog No. 502038896 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 10X buffer concentrate B is specifically optimized for use in creating a pH 5.0 to pH 10.2 gradient, essential for monoclonal antibody (mAb) charge variant separations. It is designed to be used alongside BioResolve SCX columns and features a pH of 10.2.
  • Optimized for creating a pH 5.0 to pH 10.2 gradient.
  • Aids in monoclonal antibody (mAb) charge variant separations.
  • Designed for use with BioResolve SCX columns.
  • Features a pH of 10.2.
Catalog No. 50-203-8896 Supplier Waters Corp Supplier No. 186009064
This 10X buffer concentrate B is specifically optimized for use in creating a pH 5.0 to pH 10.2 gradient, essential for monoclonal antibody (mAb) charge variant separations. It is designed to be used alongside BioResolve SCX columns and features a pH of 10.2.
  • Optimized for creating a pH 5.0 to pH 10.2 gradient.
  • Aids in monoclonal antibody (mAb) charge variant separations.
  • Designed for use with BioResolve SCX columns.
  • Features a pH of 10.2.