Learn More
Waters Corp Waters BioResolve CX pH Concentrate Kit, pH 5.0, pH 10.2
Catalog No. 502037592 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit, designed for use with BioResolve SCX columns, contains 10X buffer concentrates A and B. These concentrates are optimized for creating a pH 5.0 to 10.2 gradient, which is used for mAb charge variant separations. The kit includes one bottle of BioResolve CX pH Concentrate A (pH 5.0) and one bottle of BioResolve CX pH Concentrate B (pH 10.2).
  • Mobile phase
  • UNSPSC: 41115709
  • Application: Protein
  • Application kits
  • Units per package: 2 pk
Catalog No. 50-203-7592 Supplier Waters Corp Supplier No. 176004340
This kit, designed for use with BioResolve SCX columns, contains 10X buffer concentrates A and B. These concentrates are optimized for creating a pH 5.0 to 10.2 gradient, which is used for mAb charge variant separations. The kit includes one bottle of BioResolve CX pH Concentrate A (pH 5.0) and one bottle of BioResolve CX pH Concentrate B (pH 10.2).
  • Mobile phase
  • UNSPSC: 41115709
  • Application: Protein
  • Application kits
  • Units per package: 2 pk