Learn More
Waters Corp Waters BioResolve Guard Column, 450 Å, 2.7 μm, 2.1 mm x 30 mm
Catalog No. 502038964 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard column protects analytical columns from contaminants, extending their lifetime. It removes particulate contaminants from the mobile phase stream and is suitable for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments, operating within a pH range of 1-8.
  • Primarily used for legacy methods.
  • Provides enhanced protection from contaminants within existing workflows.
  • Can be used repeatedly.
  • Contains 2.7 μm superficially-porous-silica particles with an average pore diameter of 450 Å.
  • Functionalized with a proprietary polyphenyl bonding process for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
Catalog No. 50-203-8964 Supplier Waters Corp Supplier No. 186009270
This guard column protects analytical columns from contaminants, extending their lifetime. It removes particulate contaminants from the mobile phase stream and is suitable for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments, operating within a pH range of 1-8.
  • Primarily used for legacy methods.
  • Provides enhanced protection from contaminants within existing workflows.
  • Can be used repeatedly.
  • Contains 2.7 μm superficially-porous-silica particles with an average pore diameter of 450 Å.
  • Functionalized with a proprietary polyphenyl bonding process for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.