Learn More
Waters Corp BioResolve Protein A Affinity Column, MaxPeak Premier, 3.5 μm, 2.1 × 20 mm
Catalog No. NC3735420 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This affinity column is designed for analytical and process development scientists in PAT labs. It determines antibody titer measurements earlier in cell harvest, offering greater sensitivity for faster decision-making and improved process control. Optimized for UHPLC or UPLC conditions, this 2.1 × 20 mm column provides high sensitivity and sharp, narrow peaks for titer analysis. MaxPeak HPS technology minimizes unwanted analyte surface interactions, enhancing peak shape, recovery, and reproducibility. This aids clone selection and reduces time to downstream processes.
  • Enables earlier antibody titer determination.
  • Offers up to 5* greater sensitivity.
  • Optimized for UHPLC or UPLC conditions.
  • Reduces unwanted analyte surface interactions with MaxPeak HPS.
  • Improves peak shape, recovery, and reproducibility.
Catalog No. NC3735420 Supplier Waters Corp Supplier No. 186011369
This affinity column is designed for analytical and process development scientists in PAT labs. It determines antibody titer measurements earlier in cell harvest, offering greater sensitivity for faster decision-making and improved process control. Optimized for UHPLC or UPLC conditions, this 2.1 × 20 mm column provides high sensitivity and sharp, narrow peaks for titer analysis. MaxPeak HPS technology minimizes unwanted analyte surface interactions, enhancing peak shape, recovery, and reproducibility. This aids clone selection and reduces time to downstream processes.
  • Enables earlier antibody titer determination.
  • Offers up to 5* greater sensitivity.
  • Optimized for UHPLC or UPLC conditions.
  • Reduces unwanted analyte surface interactions with MaxPeak HPS.
  • Improves peak shape, recovery, and reproducibility.