Learn More
Waters Corp Waters Corporation Column, Reversed Phase, 2.7 μm, 450 Å, 1 mm × 50 mm
Catalog No. NC1839842 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The column is designed for reversed-phase protein separation and analysis of intact IgG monoclonal antibodies and their fragments. It features 2.7 μm superficially-porous-silica particles with a 450Å pore diameter and a proprietary polyphenyl bonding process, suitable for LC or LC-MS analyses.
  • Optimal for reverse phase protein separation.
  • Proprietary polyphenyl bonding.
  • Improved sensitivity, resolution, and throughput.
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Enhanced sample loading.
  • Minimized injection-to-injection carryover.
  • Rigorous QC process with NIST-based mAb standards.
  • Protection against particulate contaminants.
Catalog No. NC1839842 Supplier Waters Corp Supplier No. 186009015
The column is designed for reversed-phase protein separation and analysis of intact IgG monoclonal antibodies and their fragments. It features 2.7 μm superficially-porous-silica particles with a 450Å pore diameter and a proprietary polyphenyl bonding process, suitable for LC or LC-MS analyses.
  • Optimal for reverse phase protein separation.
  • Proprietary polyphenyl bonding.
  • Improved sensitivity, resolution, and throughput.
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Enhanced sample loading.
  • Minimized injection-to-injection carryover.
  • Rigorous QC process with NIST-based mAb standards.
  • Protection against particulate contaminants.