Learn More
Waters Corp BioResolve RP mAb Polyphenyl Method Validation Kit, 450Å, 2.7 μm, 3/pk
Catalog No. 502390811 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Method Validation Kit simplifies method validation for LC-MS separations. It includes three columns from different batches, along with standards to ensure system performance and facilitate data verification through sequential separations.
  • Columns contain 2.7-μm superficially-porous-silica particles (450Å pore diameter).
  • Functionalized with a proprietary polyphenyl bonding process for intact IgG mAb and fragment analysis.
  • Kit includes three columns (same sorbent, different batches) and standards (Intact mAb Mass Check, mAb Subunit).
  • Novel phenyl bonding improves peak capacity for protein analysis (mAbs, ADCs).
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Minimal injection-to-injection carryover; operates at lower temperatures for sample integrity.
Catalog No. 50-239-0811 Supplier Waters Corp Supplier No. 176004160
The BioResolve RP mAb Polyphenyl Method Validation Kit simplifies method validation for LC-MS separations. It includes three columns from different batches, along with standards to ensure system performance and facilitate data verification through sequential separations.
  • Columns contain 2.7-μm superficially-porous-silica particles (450Å pore diameter).
  • Functionalized with a proprietary polyphenyl bonding process for intact IgG mAb and fragment analysis.
  • Kit includes three columns (same sorbent, different batches) and standards (Intact mAb Mass Check, mAb Subunit).
  • Novel phenyl bonding improves peak capacity for protein analysis (mAbs, ADCs).
  • Compatible with UHPLC, UPLC, and HPLC systems.
  • Minimal injection-to-injection carryover; operates at lower temperatures for sample integrity.