Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp BioResolve Method Validation Kit, 450Å, 2.7 μm, 2.1 mm X 150 mm, 3 pk

Catalog No. 502390635 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The BioResolve RP mAb Polyphenyl Method Validation Kit is designed for reverse phase protein separation and analysis, specifically for intact IgG monoclonal antibodies and their fragments using LC or LC-MS analyses. This kit includes three columns from different manufactured batches, which are essential for method development and validating data reproducibility.

  • Separation mode: Reversed phase
  • Temperature limits: 90°C
  • Maximum pressure: 10000 psi (689 Bar)
  • Particle size: 2.7 μm
  • Pore size: 450 Å
  • System compatibility: HPLC, UHPLC, UPLC
  • Composed of 2.7 μm superficially-porous-silica particles with a proprietary polyphenyl bonding process.
  • Kit contains three columns from three different manufactured batches.
  • High-coverage bonding increases low pH stability, offering flexibility for method development.

Catalog No. 50-239-0635 Supplier Waters Corp Supplier No. 186008957
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The BioResolve RP mAb Polyphenyl Method Validation Kit is designed for reverse phase protein separation and analysis, specifically for intact IgG monoclonal antibodies and their fragments using LC or LC-MS analyses. This kit includes three columns from different manufactured batches, which are essential for method development and validating data reproducibility.

  • Separation mode: Reversed phase
  • Temperature limits: 90°C
  • Maximum pressure: 10000 psi (689 Bar)
  • Particle size: 2.7 μm
  • Pore size: 450 Å
  • System compatibility: HPLC, UHPLC, UPLC
  • Composed of 2.7 μm superficially-porous-silica particles with a proprietary polyphenyl bonding process.
  • Kit contains three columns from three different manufactured batches.
  • High-coverage bonding increases low pH stability, offering flexibility for method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.