Learn More
Waters Corp BioResolve RP mAb Polyphenyl Kit, 450Å, 2.7μm
Catalog No. 502390792 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit is designed for monoclonal antibody analysis, streamlining LC-MS method development and validation. It contains three columns with 2.7 μm superficially-porous-silica particles and 450 Å pore diameter, optimized for intact IgG monoclonal antibodies and their fragments. The kit includes Intact mAb Mass Check Standard and mAb Subunit Standard for system performance verification.
  • Dedicated to data integrity.
  • Enhanced performance from three different batches of column material.
  • Strict QC testing with Intact mAb Mass Check and mAb Subunit Standards.
  • Optimized for mAb analysis using a novel phenyl bonding process.
  • Improved peak capacity for better component resolution.
  • Includes standards for system performance verification.
  • eCord Connector for convenience.
  • Recommended VanGuard Cartridge for contamination protection.
Catalog No. 50-239-0792 Supplier Waters Corp Supplier No. 176004165
This Method Validation Kit is designed for monoclonal antibody analysis, streamlining LC-MS method development and validation. It contains three columns with 2.7 μm superficially-porous-silica particles and 450 Å pore diameter, optimized for intact IgG monoclonal antibodies and their fragments. The kit includes Intact mAb Mass Check Standard and mAb Subunit Standard for system performance verification.
  • Dedicated to data integrity.
  • Enhanced performance from three different batches of column material.
  • Strict QC testing with Intact mAb Mass Check and mAb Subunit Standards.
  • Optimized for mAb analysis using a novel phenyl bonding process.
  • Improved peak capacity for better component resolution.
  • Includes standards for system performance verification.
  • eCord Connector for convenience.
  • Recommended VanGuard Cartridge for contamination protection.