Learn More
Waters Corp RP mAb Polyphenyl Column, 450Å, 2.7 μm, 3 mm x 150 mm
Catalog No. 502391034 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These RP mAb columns feature 2.7 μm superficially-porous-silica particles with an average pore diameter of 450 Å. Functionalized with a proprietary polyphenyl bonding process, they are designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. The column helps overcome analytical challenges, offering better resolution at higher flow rates, beneficial for method development.
  • Proprietary polyphenyl bonding process on superficially-porous-particles.
  • Better peak capacity for improved component resolution.
  • Minimal injection-to-injection carryover.
  • Wide and usable pH range.
  • Includes Intact mAb Mass Check Standard.
  • Includes mAb Subunit Standard.
  • Undergoes strict QC testing.
  • VanGuard Cartridge option available.
Catalog No. 50-239-1034 Supplier Waters Corp Supplier No. 176004164
These RP mAb columns feature 2.7 μm superficially-porous-silica particles with an average pore diameter of 450 Å. Functionalized with a proprietary polyphenyl bonding process, they are designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. The column helps overcome analytical challenges, offering better resolution at higher flow rates, beneficial for method development.
  • Proprietary polyphenyl bonding process on superficially-porous-particles.
  • Better peak capacity for improved component resolution.
  • Minimal injection-to-injection carryover.
  • Wide and usable pH range.
  • Includes Intact mAb Mass Check Standard.
  • Includes mAb Subunit Standard.
  • Undergoes strict QC testing.
  • VanGuard Cartridge option available.