Learn More
Waters Corp BioResolve RP mAb Polyphenyl Method Validation Kit, 450Å, 2.7 μm, 1 mm × 150 mm
Catalog No. 502390787 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Method Validation Kit is designed to streamline purchasing and enable rapid reproduction and preparation of samples for method transfer in intact protein analysis of monoclonal antibodies and ADCs.
  • Includes three columns from different manufactured batches for method validation.
  • Includes Intact mAb Mass Check Standard and mAb Subunit Standard to verify LC-MS or MS system performance.
  • Columns feature 2.7-μm superficially-porous-silica particles with a 450Å pore diameter and proprietary polyphenyl bonding.
  • Flexible for use on all UHPLC, UPLC, and HPLC systems.
  • Withstands difficult mAb analysis, tolerating up to 90°C (194°F) and a broad pH range.
Catalog No. 50-239-0787 Supplier Waters Corp Supplier No. 176004289
The BioResolve RP mAb Polyphenyl Method Validation Kit is designed to streamline purchasing and enable rapid reproduction and preparation of samples for method transfer in intact protein analysis of monoclonal antibodies and ADCs.
  • Includes three columns from different manufactured batches for method validation.
  • Includes Intact mAb Mass Check Standard and mAb Subunit Standard to verify LC-MS or MS system performance.
  • Columns feature 2.7-μm superficially-porous-silica particles with a 450Å pore diameter and proprietary polyphenyl bonding.
  • Flexible for use on all UHPLC, UPLC, and HPLC systems.
  • Withstands difficult mAb analysis, tolerating up to 90°C (194°F) and a broad pH range.