Learn More
Waters Corp BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 3 mm X 100 mm
Catalog No. 502390817 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioResolve RP mAb Columns provide an innovative solution for characterization of proteins using reverse-phase chromatography. These columns are designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Contains 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å.
  • Functionalized with a proprietary polyphenyl bonding process.
  • Utilizes novel phenyl bonding and solid-core technology for improved chromatographic resolution at higher flow rates.
  • Includes Intact mAb Mass Check Standard and mAb Subunit Standard.
  • Undergoes strict QC testing with included standards to ensure proper performance.
  • Can be protected from contaminants with VanGuard column protection technology.
Catalog No. 50-239-0817 Supplier Waters Corp Supplier No. 176004163
BioResolve RP mAb Columns provide an innovative solution for characterization of proteins using reverse-phase chromatography. These columns are designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Contains 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å.
  • Functionalized with a proprietary polyphenyl bonding process.
  • Utilizes novel phenyl bonding and solid-core technology for improved chromatographic resolution at higher flow rates.
  • Includes Intact mAb Mass Check Standard and mAb Subunit Standard.
  • Undergoes strict QC testing with included standards to ensure proper performance.
  • Can be protected from contaminants with VanGuard column protection technology.