Learn More
Waters Corp BioResolve RP mAb Polyphenyl Method Validation Kit, 450 Å, 2.7 μm
Catalog No. 502391249 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Method Validation Kit is for LC and LC-MS analyses of intact IgG monoclonal antibodies and their fragments. It uses columns with 2.7-μm solid-core silica particles and 450 Å pore diameter, with a proprietary polyphenyl bonding process. This kit enhances performance by minimizing analyte surface interactions, improving peak shape, recovery, and reproducibility.
  • Columns feature 2.7-μm solid-core silica particles (450 Å average pore diameter).
  • Proprietary polyphenyl bonding process for mAb analysis.
  • MaxPeak HPS minimizes analyte-surface interactions.
  • Compatible with HPLC, UHPLC, and UPLC™.
  • eCord™ stores column performance history and usage data.
  • Tolerates 10,000 psi (689 bar) from 50°C to 90°C with pH 2 to 7.
Catalog No. 50-239-1249 Supplier Waters Corp Supplier No. 186009019
The BioResolve RP mAb Polyphenyl Method Validation Kit is for LC and LC-MS analyses of intact IgG monoclonal antibodies and their fragments. It uses columns with 2.7-μm solid-core silica particles and 450 Å pore diameter, with a proprietary polyphenyl bonding process. This kit enhances performance by minimizing analyte surface interactions, improving peak shape, recovery, and reproducibility.
  • Columns feature 2.7-μm solid-core silica particles (450 Å average pore diameter).
  • Proprietary polyphenyl bonding process for mAb analysis.
  • MaxPeak HPS minimizes analyte-surface interactions.
  • Compatible with HPLC, UHPLC, and UPLC™.
  • eCord™ stores column performance history and usage data.
  • Tolerates 10,000 psi (689 bar) from 50°C to 90°C with pH 2 to 7.