Learn More
Waters Corp BioResolve RP mAb Polyphenyl Method Validation Kit, 450Å, 2.7 μm
Catalog No. 502391215 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Method Validation Kit is designed for reverse phase protein separation and analysis of intact IgG monoclonal antibodies and their fragments, compatible with all UHPLC, UPLC, and HPLC systems.
  • Method validation kit: contains three columns from different manufactured batches for method development and validation.
  • Proprietary polyphenyl bonding: specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Rugged silica particles: offers stability even at low pH.
  • High coverage bonding: actively prevents carryover effects during method development.
  • System compatibility: can be used on all UHPLC, UPLC, and HPLC systems.
  • Column protection: compatible with VanGuard cartridge for extended column lifetime.
  • Quality control: each batch is qualified using a gradient separation of the mAb subunit standard.
Catalog No. 50-239-1215 Supplier Waters Corp Supplier No. 186009018
The BioResolve RP mAb Polyphenyl Method Validation Kit is designed for reverse phase protein separation and analysis of intact IgG monoclonal antibodies and their fragments, compatible with all UHPLC, UPLC, and HPLC systems.
  • Method validation kit: contains three columns from different manufactured batches for method development and validation.
  • Proprietary polyphenyl bonding: specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Rugged silica particles: offers stability even at low pH.
  • High coverage bonding: actively prevents carryover effects during method development.
  • System compatibility: can be used on all UHPLC, UPLC, and HPLC systems.
  • Column protection: compatible with VanGuard cartridge for extended column lifetime.
  • Quality control: each batch is qualified using a gradient separation of the mAb subunit standard.