Learn More
Waters Corp Method Validation Kit, Reversed Phase, 2.7 μm, 450 Å
Catalog No. 502392561 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit has 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å. Functionalized with a proprietary polyphenyl bonding, it is ideal for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. Each kit includes three columns from three different manufactured batches.
  • Designed to provide a reversed-phase solution for intact and IdeS digested monoclonal antibodies.
  • Novel polyphenyl bonding on superficially porous silica particles is specific for LC-MS or LC analysis of intact IgG monoclonal antibodies.
  • High-coverage bonding prevents carryover effects and improves low pH stability.
  • Included columns are compatible with UHPLC, UPLC, and HPLC systems, offering method transfer flexibility.
  • Phenyl bonding and solid-core technology maximize resolving power and selectivity for protein sample analysis.
Catalog No. 50-239-2561 Supplier Waters Corp Supplier No. 186008959
This method validation kit has 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å. Functionalized with a proprietary polyphenyl bonding, it is ideal for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. Each kit includes three columns from three different manufactured batches.
  • Designed to provide a reversed-phase solution for intact and IdeS digested monoclonal antibodies.
  • Novel polyphenyl bonding on superficially porous silica particles is specific for LC-MS or LC analysis of intact IgG monoclonal antibodies.
  • High-coverage bonding prevents carryover effects and improves low pH stability.
  • Included columns are compatible with UHPLC, UPLC, and HPLC systems, offering method transfer flexibility.
  • Phenyl bonding and solid-core technology maximize resolving power and selectivity for protein sample analysis.