Learn More
Waters Corp Waters BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 4.6 mm x 50 mm
Catalog No. 501528574 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioResolve RP mAb Columns contain 2.7 μm superficially-porous-silica particles with an average pore diameter of 450 Å. Functionalized with a proprietary polyphenyl bonding process, these columns are designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Improved peak capacity for better component resolution.
  • Wide, usable pH range to accommodate varying mobile phases.
  • Equipped with VanGuard column protection technology to prevent data variability caused by particulate contaminants.
  • Novel phenyl bonding process for characterization of intact protein analysis of mAbs and ADCs, providing better resolution at higher flow rates.
  • Compatible with UHPLC, UPLC, and HPLC systems for method development and transfer.
Catalog No. 50-152-8574 Supplier Waters Corp Supplier No. 176004167
BioResolve RP mAb Columns contain 2.7 μm superficially-porous-silica particles with an average pore diameter of 450 Å. Functionalized with a proprietary polyphenyl bonding process, these columns are designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Improved peak capacity for better component resolution.
  • Wide, usable pH range to accommodate varying mobile phases.
  • Equipped with VanGuard column protection technology to prevent data variability caused by particulate contaminants.
  • Novel phenyl bonding process for characterization of intact protein analysis of mAbs and ADCs, providing better resolution at higher flow rates.
  • Compatible with UHPLC, UPLC, and HPLC systems for method development and transfer.