Learn More
Waters Corp Waters RP mAb Polyphenyl Column, 450Å, 2.7 μm
Supplier:  Waters Corp 186008945
Encompass_Preferred
The BioResolve RP mAb Polyphenyl Column is designed for reverse phase protein separation and analysis. It features 2.7 μm superficially-porous-silica particles with 450Å pore diameter, enhanced by a proprietary polyphenyl bonding process for LC/LC-MS analysis of intact IgG monoclonal antibodies and their fragments.
  • Offers enhanced peak capacity and component resolution.
  • Ensures low injection-to-injection carryover.
  • Permits lower temperature operation to reduce sample degradation.
  • Compatible with UPLC, UHPLC, and HPLC platforms.
  • Manufactured in a cGMP, ISO 9001 certified facility.
  • Includes eCord with Certificate of Analysis and Performance Test Chromatogram.
  • Tolerates pressures up to 10,000 psi (689 bar).
Catalog No. 50-152-8592