Learn More
Waters Corp Waters BioResolve RP mAb Polyphenyl Column, 450Å, 2.7 μm, 4.6 mm X 50 mm
Catalog No. 501528595 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is the first choice for reverse phase protein separation and analysis. It contains 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å. The particles are functionalized with a proprietary polyphenyl bonding process specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. Each batch is qualified using a gradient separation of a mAb Subunit Standard.
  • Designed for use on all UHPLC, UPLC, or HPLC systems with minimal injection-to-injection carryover.
  • The combination of the bonding process and solid-core column technology allows for better resolution at higher flow rates, aiding method transfer from HPLC to UPLC systems.
  • High-coverage bonding prevents carryover effects and enhances low pH stability, contributing to longer column lifetimes.
Catalog No. 50-152-8595 Supplier Waters Corp Supplier No. 186008953
Add to Cart
Add to Cart
This column is the first choice for reverse phase protein separation and analysis. It contains 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å. The particles are functionalized with a proprietary polyphenyl bonding process specifically designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. Each batch is qualified using a gradient separation of a mAb Subunit Standard.
  • Designed for use on all UHPLC, UPLC, or HPLC systems with minimal injection-to-injection carryover.
  • The combination of the bonding process and solid-core column technology allows for better resolution at higher flow rates, aiding method transfer from HPLC to UPLC systems.
  • High-coverage bonding prevents carryover effects and enhances low pH stability, contributing to longer column lifetimes.