Learn More
Waters Corp BioResolve Column, Polyphenyl, 450 Å, 2.7 μm, 2.1 mm x 20 mm
Catalog No. NC3254105 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This BioResolve Premier RP mAb Polyphenyl Column is designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. It features 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å and a proprietary polyphenyl bonding process. The column hardware integrates MaxPeak High Performance Surface (HPS) to minimize unwanted analyte surface interactions, enhancing peak shape, recovery, and reproducibility for high-throughput Reversed-phase (RP) chromatography.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 7
  • 2.7 μm particle size
  • 450 Å pore size
  • Compatible with HPLC, UHPLC, and UPLC systems
  • Suitable for LC/MS technique
  • 2.1 mm inner diameter
  • 20 mm length
Catalog No. NC3254105 Supplier Waters Corp Supplier No. 186011019
This BioResolve Premier RP mAb Polyphenyl Column is designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments. It features 2.7 μm superficially-porous-silica particles with an average pore diameter of 450Å and a proprietary polyphenyl bonding process. The column hardware integrates MaxPeak High Performance Surface (HPS) to minimize unwanted analyte surface interactions, enhancing peak shape, recovery, and reproducibility for high-throughput Reversed-phase (RP) chromatography.
  • Phenyl chemistry
  • Reversed phase separation mode
  • Silica particle substrate
  • pH range of 2 to 7
  • 2.7 μm particle size
  • 450 Å pore size
  • Compatible with HPLC, UHPLC, and UPLC systems
  • Suitable for LC/MS technique
  • 2.1 mm inner diameter
  • 20 mm length