Learn More
Waters Corp WATERS CORPORATION BioResolve Method Validation Kit, 450Å, 2.7 μm
Catalog No. 501528578 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The BioResolve RP mAb Columns feature 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å. These particles are functionalized with a proprietary polyphenyl bonding process designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Separation mode: Reverse phase
  • Particle size: 2.7 μm
  • Pore size: 450 Å
  • Units per package: 3 pk
  • Includes three columns from three different manufactured batches
  • Includes three vials of the Intact mAb Mass Check Standard
  • Includes three vials of the mAb Subunit Standard
  • Designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments
Catalog No. 50-152-8578 Supplier Waters Corp Supplier No. 176004171
The BioResolve RP mAb Columns feature 2.7-μm superficially-porous-silica particles with an average pore diameter of 450Å. These particles are functionalized with a proprietary polyphenyl bonding process designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments.
  • Separation mode: Reverse phase
  • Particle size: 2.7 μm
  • Pore size: 450 Å
  • Units per package: 3 pk
  • Includes three columns from three different manufactured batches
  • Includes three vials of the Intact mAb Mass Check Standard
  • Includes three vials of the mAb Subunit Standard
  • Designed for LC or LC-MS analyses of intact IgG monoclonal antibodies and their fragments