Learn More
Waters Corp Waters Method Validation Kit, BioResolve RP mAb Polyphenyl, 450 Å
Catalog No. 501528598 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit provides a reverse-phase solution for protein separation and analysis, ideal for intact IgG monoclonal antibodies and their fragments. It contains three BioResolve RP mAb Polyphenyl Columns with 2.7 μm superficially-porous-silica particles and a 450Å average pore diameter. The columns utilize solid-core technology and a proprietary polyphenyl bonding process to deliver enhanced chromatographic resolution and selectivity at high flow rates.
  • Includes three columns with 2.7 μm superficially-porous-silica particles and 450Å average pore diameter
  • Features proprietary polyphenyl bonding process for LC or LC-MS analysis of intact IgG monoclonal antibodies and their fragments
  • Utilizes solid-core column technology to enhance chromatographic resolution and selectivity
  • Undergoes a strict QC process with NIST-based mAb standards for optimal performance
Catalog No. 50-152-8598 Supplier Waters Corp Supplier No. 186008956
This kit provides a reverse-phase solution for protein separation and analysis, ideal for intact IgG monoclonal antibodies and their fragments. It contains three BioResolve RP mAb Polyphenyl Columns with 2.7 μm superficially-porous-silica particles and a 450Å average pore diameter. The columns utilize solid-core technology and a proprietary polyphenyl bonding process to deliver enhanced chromatographic resolution and selectivity at high flow rates.
  • Includes three columns with 2.7 μm superficially-porous-silica particles and 450Å average pore diameter
  • Features proprietary polyphenyl bonding process for LC or LC-MS analysis of intact IgG monoclonal antibodies and their fragments
  • Utilizes solid-core column technology to enhance chromatographic resolution and selectivity
  • Undergoes a strict QC process with NIST-based mAb standards for optimal performance