Learn More
Waters Corp BIORESOLVE RP MAB POLYPHENYL M
Catalog No. 501528600 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
BioResolve RP mAb Polyphenyl Method Validation Kit, 450A, 2.7 um, 4.6mm X 100mm, 3/pkg;Silica, 1 - 8 Ph Range, Low Silanol Activity, End Capped , Spherical , 5.5 % Carbon Load
Catalog No. 50-152-8600 Supplier Waters Corp Supplier No. 186008960
Add to Cart
Add to Cart
BioResolve RP mAb Polyphenyl Method Validation Kit, 450A, 2.7 um, 4.6mm X 100mm, 3/pkg;Silica, 1 - 8 Ph Range, Low Silanol Activity, End Capped , Spherical , 5.5 % Carbon Load